Assay Details
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281929.1 | NP_001268858.1 | 1 - 2 | 358 | 61 | |
NM_001290451.1 | NP_001277380.1 | 1 - 2 | 130 | 61 | ||
NM_016886.4 | NP_058582.3 | 1 - 2 | 358 | 61 | ||
NR_104053.1 | 1 - 2 | 358 | 61 | |||
GenBank mRNA | AB022342.1 | - | 1 - 2 | 148 | 61 | |
AB079072.1 | - | 1 - 2 | 94 | 61 | ||
AB102775.1 | - | 1 - 2 | 94 | 61 | ||
AB102776.1 | - | 1 - 2 | 94 | 61 | ||
AF483494.1 | - | 1 - 2 | 94 | 61 | ||
AF483495.1 | - | 1 - 2 | 94 | 61 | ||
AK135043.1 | - | 1 - 2 | 111 | 61 | ||
AK138615.1 | - | 1 - 2 | 263 | 61 | ||
AK157815.1 | - | 2 - 3 | 535 | 61 | ||
AK161163.1 | - | 1 - 2 | 178 | 61 | ||
AK220530.1 | - | 1 - 2 | 145 | 61 | ||
BC076584.1 | - | 1 - 2 | 223 | 61 | ||
BC086678.1 | - | 1 - 2 | 303 | 30668476 | 61 | |
BC129855.1 | - | 1 - 2 | 148 | 40109751 | 61 | |
BC129856.1 | - | 1 - 2 | 148 | 40109752 | 61 | |
CJ107988.1 | - | 1 - 2 | 356 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
