Assay Details
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033285.1 | NP_001028457.1 | 1 - 2 | 195 | 136 | |
GenBank mRNA | AK138649.1 | - | 1 - 2 | 526 | 136 | |
BC158017.1 | - | 1 - 2 | 195 | 9007323 | 136 | |
BC158095.1 | - | 1 - 2 | 195 | 9007382 | 136 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK138649.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
