Assay Details
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033285.1 | NP_001028457.1 | 2 - 3 | 295 | 99 | |
GenBank mRNA | AK162174.1 | - | 2 - 3 | 193 | 99 | |
BC158017.1 | - | 2 - 3 | 295 | 9007323 | 99 | |
BC158095.1 | - | 2 - 3 | 295 | 9007382 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
