Assay Details
Gene Symbol: Cdc42bpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033285.1 | NP_001028457.1 | 4 - 5 | 473 | 76 | |
GenBank mRNA | AK138649.1 | - | 5 - 6 | 962 | 76 | |
AK162174.1 | - | 4 - 5 | 371 | 76 | ||
BC158017.1 | - | 4 - 5 | 473 | 9007323 | 76 | |
BC158095.1 | - | 4 - 5 | 473 | 9007382 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
