Assay Details
Gene Symbol: Wdhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172598.3 | NP_766186.2 | 23 - 24 | 3165 | 75 | |
GenBank mRNA | AK160777.1 | - | 3 - 4 | 326 | 75 | |
BC063740.1 | - | 23 - 24 | 3098 | 6328626 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
