Assay Details
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033272.1 | NP_001028444.1 | 6 - 7 | 2659 | 68 | |
GenBank mRNA | AK151394.1 | - | 3 - 4 | 539 | 68 | |
AK220313.1 | - | 3 - 4 | 381 | 68 | ||
BC035942.1 | - | 4 - 5 | 677 | 68 | ||
BC138455.1 | - | 6 - 7 | 2616 | 8861475 | 68 | |
BC145285.1 | - | 6 - 7 | 2616 | 9053839 | 68 | |
CD352148.1 | - | 3 - 4 | 423 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
