Assay Details
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033272.1 | NP_001028444.1 | 7 - 8 | 2850 | 73 | |
GenBank mRNA | AK151394.1 | - | 4 - 5 | 730 | 73 | |
AK220313.1 | - | 4 - 5 | 572 | 73 | ||
BC035942.1 | - | 5 - 6 | 868 | 73 | ||
BC138455.1 | - | 7 - 8 | 2807 | 8861475 | 73 | |
BC145285.1 | - | 7 - 8 | 2807 | 9053839 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
