Assay Details
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033272.1 | NP_001028444.1 | 8 - 9 | 3030 | 94 | |
GenBank mRNA | AK151394.1 | - | 5 - 6 | 910 | 94 | |
AK220313.1 | - | 5 - 6 | 752 | 94 | ||
BC035942.1 | - | 6 - 7 | 1048 | 94 | ||
BC138455.1 | - | 8 - 9 | 2987 | 8861475 | 94 | |
BC145285.1 | - | 8 - 9 | 2987 | 9053839 | 94 | |
CN538317.1 | - | 2 - 3 | 212 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
