Assay Details
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033272.1 | NP_001028444.1 | 11 - 12 | 3761 | 96 | |
GenBank mRNA | AK151394.1 | - | 8 - 9 | 1643 | 96 | |
AK158193.1 | - | 3 - 4 | 1295 | 96 | ||
AK220313.1 | - | 8 - 9 | 1483 | 96 | ||
BC035942.1 | - | 9 - 10 | 1779 | 96 | ||
BC138455.1 | - | 11 - 12 | 3718 | 8861475 | 96 | |
BC145285.1 | - | 11 - 12 | 3718 | 9053839 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
