Assay Details
Gene Symbol: Tram1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146140.3 | NP_666252.1 | 1 | 1682 | 166 | |
GenBank mRNA | AK044115.1 | - | 1 | 1609 | 166 | |
AK048118.1 | - | 1632 | 166 | |||
AK083053.1 | - | 1 | 1611 | 166 | ||
BC024829.1 | - | 1 | 1609 | 5359105 | 166 | |
BC027120.1 | - | 1 | 1609 | 5359106 | 166 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492340 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492343 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492338 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492342 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492339 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492341 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | BC027120.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | BC024829.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | AK048118.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | AK083053.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | AK044115.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | BC027120.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | BC024829.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | AK048118.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | AK083053.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P144957 | AK044115.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934873 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172924 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174205 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179019 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | BC027120.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | BC024829.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | AK048118.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | AK083053.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | AK044115.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | BC027120.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | BC024829.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | AK048118.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | AK083053.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P144957 | AK044115.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492322 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492323 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492347 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492316 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492334 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492312 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492301 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492308 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492329 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492340 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492306 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492349 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492304 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492315 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492310 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492336 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492326 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492343 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492311 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492303 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492354 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492313 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492333 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492317 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492327 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22492320 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179019 | - | - |
Whole Mouse Genome G4122A | A_51_P144957 | BC027120.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | BC024829.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | AK048118.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | AK083053.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | AK044115.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | BC027120.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | BC024829.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | AK048118.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | AK083053.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P144957 | AK044115.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133360 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179019 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02491746 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02363721 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02491740 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02363731 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492339 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492343 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492342 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492340 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492341 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22492338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22492308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02363731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02613475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02491747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02428305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02363711 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02363718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492290 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02363721 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492396 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02363732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492335 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492311 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492304 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02544929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492393 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492333 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02363730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492301 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02970306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02769111 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492312 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492360 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22492376 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10495870 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10495871 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451268_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10495869 | - | - |
Mouse Genome 430 2.0 Array | 1451268_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451268_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229801_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229801_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300010293.mm.1 | TC0300001260.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300010293.mm.1 | NM_146140.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300010293.mm.1 | NM_146140.3 | Refseq |
Mouse Expression Set 430 | 1451268_PM_at | - | - |
Mouse Expression Set 430 | 1451268_at | - | - |
Murine Genome U74v2 Set | 135813_PM_at | - | - |
Murine Genome U74v2 Set | 166275_PM_i_at | - | - |
Murine Genome U74v2 Set | 135813_at | - | - |
Murine Genome U74v2 Set | 166275_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1451268_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451268_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451268_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451268_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 910149 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 50128 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106350017 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 50128 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000050128 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2484215 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237853 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2484215 | - | - |
mouse-6 v1.1 (Illumina) | scl00229801.2_16-S | - | - |
mouse-6 v1.1 (Illumina) | scl22656.1_111-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106350017 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 50128 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00229801.2_16-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 50128 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106350017 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2484215 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2484215 | - | - |
