Assay Details
Gene Symbol: C1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183175.4 | NP_898998.2 | 4 | 1252 | 102 | |
GenBank mRNA | AK033673.1 | - | 1 | 937 | 102 | |
DQ002401.1 | - | 4 | 1252 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P200610 | DQ002401.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P200610 | NM_183175.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P200610 | AK033673.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P200610 | DQ002401.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P200610 | NM_183175.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P200610 | AK033673.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45721 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606561 | AK049904.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804560 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528300 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102367 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637257 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3289 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172113 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P200610 | DQ002401.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P200610 | NM_183175.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P200610 | AK033673.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P200610 | DQ002401.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P200610 | NM_183175.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P200610 | AK033673.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29646465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29646459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29646461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29646462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29646460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29646463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29646466 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172113 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08131108 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08131126 | - | - |
Whole Mouse Genome G4122A | A_51_P127475 | DQ002401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P127475 | NM_183175.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P127475 | AK033673.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200610 | DQ002401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200610 | NM_183175.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200610 | AK033673.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200610 | DQ002401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200610 | NM_183175.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200610 | AK033673.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134838 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646411 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29646381 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11769716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646363 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646386 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646405 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646353 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646414 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11639690 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646429 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646342 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11734296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11434483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11633293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646381 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646418 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11633301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646367 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646370 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646336 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11779496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11633282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11523047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11559562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11762890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646460 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646400 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11559550 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11633277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646465 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646343 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11523038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646403 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11468234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646420 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11493450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646376 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646397 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11434486 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646380 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646351 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11633300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11559557 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11769720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646463 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646372 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29646383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11613348 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10415740 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415739 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415741 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10415738 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1442033_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10415737 | - | - |
Mouse Genome 430 2.0 Array | 1442033_at | - | - |
Mouse Genome 430 2.0 Array | 1442033_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 239126_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 239126_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400006968.mm.1 | TC1400000853.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400006968.mm.1 | NM_183175.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400006968.mm.1 | NM_183175.4 | Refseq |
Mouse Expression Set 430 | 1442033_PM_at | - | - |
Mouse Expression Set 430 | 1442033_at | - | - |
Murine Genome U74v2 Set | 140572_PM_at | - | - |
Murine Genome U74v2 Set | 140572_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442033_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1442033_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 527520 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2370154 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002370154 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2716968 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2716968 | - | - |
mouse-6 v1.1 (Illumina) | scl0239126.1_315-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2370154 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0239126.1_315-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370154 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2716968 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2716968 | - | - |
