Assay Details
Gene Symbol: Adcy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080435.1 | NP_536683.1 | 14 - 15 | 1830 | 89 | |
GenBank mRNA | AF442771.1 | - | 14 - 15 | 1830 | 89 | |
AK141767.1 | - | 14 - 15 | 1901 | 89 | ||
AK161880.1 | - | 14 - 15 | 1849 | 89 | ||
AK220443.1 | - | 11 - 12 | 1953 | 89 | ||
BC015299.1 | - | 14 - 15 | 1830 | 4236007 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
