Assay Details
Gene Symbol: Nfatc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168346.1 | NP_001161818.1 | 4 - 5 | 1797 | 62 | |
NM_023699.3 | NP_076188.3 | 4 - 5 | 1797 | 62 | ||
GenBank mRNA | AK159078.1 | - | 4 - 5 | 1643 | 62 | |
EU887656.1 | - | 4 - 5 | 1555 | 62 | ||
EU887657.1 | - | 4 - 5 | 1555 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
