Assay Details
Gene Symbol: Nfatc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168346.1 | NP_001161818.1 | 7 - 8 | 2170 | 81 | |
NM_023699.3 | NP_076188.3 | 7 - 8 | 2170 | 81 | ||
GenBank mRNA | AK159078.1 | - | 7 - 8 | 2016 | 81 | |
EU887656.1 | - | 7 - 8 | 1928 | 81 | ||
EU887657.1 | - | 7 - 8 | 1928 | 81 | ||
EU887658.1 | - | 2 - 3 | 383 | 81 | ||
EU887659.1 | - | 2 - 3 | 383 | 81 | ||
EU887660.1 | - | 1 - 2 | 1439 | 81 | ||
EU887661.1 | - | 1 - 2 | 1439 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the EU887658.1, EU887659.1, EU887660.1, EU887661.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
