Assay Details
Gene Symbol: Nfatc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023699.3 | NP_076188.3 | 9 - 10 | 2881 | 87 | |
GenBank mRNA | AK159078.1 | - | 9 - 10 | 2727 | 87 | |
AW493556.1 | - | 1 - 2 | 87 | |||
BC028928.1 | - | 9 - 10 | 2668 | 3670623 | 87 | |
EU887656.1 | - | 9 - 10 | 2639 | 87 | ||
EU887658.1 | - | 4 - 5 | 1094 | 87 | ||
EU887660.1 | - | 3 - 4 | 2150 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
