Assay Details
Gene Symbol: Il16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010551.3 | NP_034681.2 | 6 - 7 | 978 | 121 | |
GenBank mRNA | AF175292.1 | - | 6 - 7 | 978 | 121 | |
AK137237.1 | - | 7 - 8 | 1611 | 121 | ||
BC058709.1 | - | 6 - 7 | 1011 | 6841305 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
