Assay Details
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172525.2 | NP_766113.1 | 15 - 16 | 1796 | 81 | |
GenBank mRNA | AK043637.1 | - | 15 - 16 | 1843 | 81 | |
AK044979.1 | - | 15 - 16 | 1846 | 81 | ||
AK132716.1 | - | 10 - 11 | 1182 | 81 | ||
BC031572.1 | - | 4 - 5 | 463 | 81 | ||
BC040387.1 | - | 15 - 16 | 1820 | 3599962 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
