Assay Details
Gene Symbol: Ltk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008523.2 | NP_032549.2 | 4 - 5 | 628 | 66 | |
NM_203345.2 | NP_976220.2 | 10 - 11 | 1474 | 66 | ||
NM_206941.1 | NP_996824.1 | 5 - 6 | 811 | 66 | ||
NM_206942.2 | NP_996825.2 | 9 - 10 | 1291 | 66 | ||
GenBank mRNA | AK157478.1 | - | 3 - 4 | 442 | 66 | |
BC047138.1 | - | 4 - 5 | 628 | 6389834 | 66 | |
BC055898.1 | - | 4 - 5 | 622 | 66 | ||
BC141299.1 | - | 9 - 10 | 1267 | 9055911 | 66 | |
M90470.1 | - | 10 - 11 | 1474 | 66 | ||
X07984.1 | - | 4 - 5 | 602 | 66 | ||
X52621.1 | - | 4 - 5 | 617 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
