Assay Details
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176942.4 | NP_795916.1 | 2 - 3 | 425 | 87 | |
GenBank mRNA | AK038476.1 | - | 2 - 3 | 427 | 87 | |
AK083185.1 | - | 2 - 3 | 212 | 87 | ||
BC062112.1 | - | 2 - 3 | 423 | 6815819 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
