Assay Details
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176942.4 | NP_795916.1 | 3 - 4 | 548 | 64 | |
GenBank mRNA | AF090374.1 | - | 1 - 2 | 113 | 64 | |
AK038476.1 | - | 3 - 4 | 550 | 64 | ||
AK083185.1 | - | 3 - 4 | 335 | 64 | ||
BC062112.1 | - | 3 - 4 | 546 | 6815819 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
