Assay Details
Gene Symbol: Fam208b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134063.3 | NP_598824.3 | 16 - 17 | 7300 | 109 | |
GenBank mRNA | AK128934.1 | - | 3 - 4 | 2617 | 109 | |
AK131707.1 | - | 5 - 6 | 4696 | 109 | ||
AK220437.1 | - | 16 - 17 | 7288 | 109 | ||
BC016423.1 | - | 3 - 4 | 1984 | 109 | ||
BC045617.1 | - | 3 - 4 | 2323 | 109 | ||
BC085160.1 | - | 7 - 8 | 6043 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
