Assay Details
Gene Symbol: Aloxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011786.2 | NP_035916.2 | 13 - 14 | 1811 | 57 | |
GenBank mRNA | BC116258.1 | - | 13 - 14 | 1810 | 40093733 | 57 |
BC116259.1 | - | 13 - 14 | 1810 | 40093735 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
