Assay Details
Gene Symbol: Acsl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033600.1 | NP_001028772.1 | 1 - 2 | 233 | 136 | |
NM_019477.3 | NP_062350.3 | 1 - 2 | 233 | 139 | ||
GenBank mRNA | AB033885.1 | - | 1 - 2 | 42 | 139 | |
AJ243502.1 | - | 1 - 2 | 81 | 136 | ||
AK054387.1 | - | 1 - 2 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB033885.1, AJ243502.1, AK054387.1, NM_001033600.1, NM_019477.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
