Assay Details
Gene Symbol: Gucy2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007576.2 | NP_001007577.1 | 10 - 11 | 2374 | 99 | |
GenBank mRNA | AK044234.1 | - | 10 - 11 | 2375 | 99 | |
AY651761.1 | - | 10 - 11 | 2369 | 99 | ||
BC115714.1 | - | 9 - 10 | 2204 | 40085764 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
