Assay Details
Gene Symbol: Mpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164733.1 | NP_001158205.1 | 6 - 7 | 928 | 143 | |
NM_001164734.1 | NP_001158206.1 | 6 - 7 | 938 | 143 | ||
GenBank mRNA | AF199010.1 | - | 6 - 7 | 934 | 143 | |
AK144434.1 | - | 6 - 7 | 939 | 143 | ||
AK164549.1 | - | 6 - 7 | 929 | 143 | ||
BC145363.1 | - | 6 - 7 | 796 | 9053921 | 143 | |
BC145364.1 | - | 6 - 7 | 786 | 9053922 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
