Assay Details
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018773.2 | NP_061243.1 | 11 - 12 | 1126 | 70 | |
GenBank mRNA | AB014485.1 | - | 11 - 12 | 1043 | 70 | |
AF051324.1 | - | 11 - 12 | 1041 | 70 | ||
AK076000.1 | - | 11 - 12 | 1071 | 70 | ||
AK144289.1 | - | 11 - 12 | 1063 | 70 | ||
BC003711.1 | - | 11 - 12 | 1126 | 3599914 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
