Assay Details
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163640.1 | NP_001157112.1 | 10 - 11 | 1160 | 97 | |
NM_023543.2 | NP_076032.2 | 4 - 5 | 663 | 97 | ||
GenBank mRNA | AJ279014.1 | - | 5 - 6 | 561 | 97 | |
AK006398.1 | - | 5 - 6 | 757 | 97 | ||
AK131641.1 | - | 10 - 11 | 1054 | 97 | ||
AK160255.1 | - | 5 - 6 | 492 | 97 | ||
BC051139.1 | - | 4 - 5 | 664 | 6589539 | 97 | |
BC141060.1 | - | 4 - 5 | 622 | 9055672 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
