Assay Details
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163640.1 | NP_001157112.1 | 2 - 3 | 268 | 85 | |
GenBank mRNA | AK030336.1 | - | 2 - 3 | 269 | 85 | |
AK131641.1 | - | 2 - 3 | 162 | 85 | ||
BB617538.1 | - | 2 - 3 | 269 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
