Assay Details
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163640.1 | NP_001157112.1 | 5 - 6 | 466 | 60 | |
GenBank mRNA | AK030336.1 | - | 5 - 6 | 467 | 60 | |
AK131641.1 | - | 5 - 6 | 360 | 60 | ||
BB617538.1 | - | 5 - 6 | 467 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
