Assay Details
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163640.1 | NP_001157112.1 | 7 - 8 | 836 | 85 | |
NM_023543.2 | NP_076032.2 | 1 - 2 | 339 | 85 | ||
GenBank mRNA | AJ279014.1 | - | 2 - 3 | 237 | 85 | |
AK006398.1 | - | 2 - 3 | 433 | 85 | ||
AK131641.1 | - | 7 - 8 | 730 | 85 | ||
AK160255.1 | - | 2 - 3 | 168 | 85 | ||
BC051139.1 | - | 1 - 2 | 340 | 6589539 | 85 | |
BC141060.1 | - | 1 - 2 | 298 | 9055672 | 85 | |
CX238009.1 | - | 1 - 2 | 339 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
