Assay Details
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163640.1 | NP_001157112.1 | 8 - 9 | 909 | 93 | |
NM_023543.2 | NP_076032.2 | 2 - 3 | 412 | 93 | ||
GenBank mRNA | AJ279014.1 | - | 3 - 4 | 310 | 93 | |
AK006398.1 | - | 3 - 4 | 506 | 93 | ||
AK131641.1 | - | 8 - 9 | 803 | 93 | ||
AK160255.1 | - | 3 - 4 | 241 | 93 | ||
BC051139.1 | - | 2 - 3 | 413 | 6589539 | 93 | |
BC141060.1 | - | 2 - 3 | 371 | 9055672 | 93 | |
CX238009.1 | - | 2 - 3 | 412 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
