Assay Details
Gene Symbol: Adamtsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029981.1 | NP_084257.1 | 8 - 9 | 952 | 89 | |
GenBank mRNA | AB096928.1 | - | 8 - 9 | 952 | 89 | |
AK156991.1 | - | 8 - 9 | 955 | 89 | ||
BC128568.1 | - | 8 - 9 | 904 | 40097415 | 89 | |
BC129984.1 | - | 8 - 9 | 904 | 40097418 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
