Assay Details
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054081.2 | NP_473422.2 | 15 - 16 | 1720 | 112 | |
GenBank mRNA | AB794093.1 | - | 16 - 17 | 1631 | 112 | |
AB794094.1 | - | 15 - 16 | 1580 | 112 | ||
AB794095.1 | - | 16 - 17 | 1584 | 112 | ||
AB794096.1 | - | 16 - 17 | 1631 | 112 | ||
AB794097.1 | - | 15 - 16 | 1533 | 112 | ||
AB794098.1 | - | 15 - 16 | 1580 | 112 | ||
AB794099.1 | - | 15 - 16 | 1580 | 112 | ||
AB794100.1 | - | 15 - 16 | 1533 | 112 | ||
AB794101.1 | - | 16 - 17 | 1631 | 112 | ||
AB794102.1 | - | 15 - 16 | 1520 | 112 | ||
AB794103.1 | - | 15 - 16 | 1580 | 112 | ||
AF288137.1 | - | 15 - 16 | 1682 | 112 | ||
AF463504.1 | - | 16 - 17 | 1631 | 112 | ||
BC005698.1 | - | 4 - 5 | 112 | |||
BC024527.1 | - | 8 - 9 | 112 | |||
BC025167.1 | - | 4 - 5 | 112 | |||
BC112910.1 | - | 16 - 17 | 6515946 | 112 | ||
CX203022.1 | - | 3 - 4 | 416 | 112 | ||
CX213009.1 | - | 2 - 3 | 165 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
