Assay Details
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054081.2 | NP_473422.2 | 16 - 17 | 1864 | 57 | |
GenBank mRNA | AB794093.1 | - | 17 - 18 | 1775 | 57 | |
AB794094.1 | - | 16 - 17 | 1724 | 57 | ||
AB794095.1 | - | 17 - 18 | 1728 | 57 | ||
AB794097.1 | - | 16 - 17 | 1677 | 57 | ||
AB794099.1 | - | 16 - 17 | 1724 | 57 | ||
AB794100.1 | - | 16 - 17 | 1677 | 57 | ||
AB794102.1 | - | 16 - 17 | 1664 | 57 | ||
AB794103.1 | - | 16 - 17 | 1724 | 57 | ||
AF288137.1 | - | 16 - 17 | 1826 | 57 | ||
AF463504.1 | - | 17 - 18 | 1775 | 57 | ||
BC005698.1 | - | 5 - 6 | 633 | 57 | ||
BC024527.1 | - | 9 - 10 | 1090 | 57 | ||
BC025167.1 | - | 5 - 6 | 621 | 57 | ||
CX213009.1 | - | 3 - 4 | 309 | 57 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
