Assay Details
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054081.2 | NP_473422.2 | 5 - 6 | 516 | 70 | |
GenBank mRNA | AB794093.1 | - | 6 - 7 | 427 | 70 | |
AB794094.1 | - | 5 - 6 | 376 | 70 | ||
AB794095.1 | - | 6 - 7 | 427 | 70 | ||
AB794096.1 | - | 6 - 7 | 427 | 70 | ||
AB794097.1 | - | 5 - 6 | 376 | 70 | ||
AB794098.1 | - | 5 - 6 | 376 | 70 | ||
AB794099.1 | - | 5 - 6 | 376 | 70 | ||
AB794100.1 | - | 5 - 6 | 376 | 70 | ||
AB794101.1 | - | 6 - 7 | 427 | 70 | ||
AB794103.1 | - | 5 - 6 | 376 | 70 | ||
AF288137.1 | - | 5 - 6 | 478 | 70 | ||
AF463504.1 | - | 6 - 7 | 427 | 70 | ||
BC112910.1 | - | 6 - 7 | 567 | 6515946 | 70 | |
CF171555.1 | - | 3 - 4 | 289 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
