Assay Details
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054081.2 | NP_473422.2 | 7 - 8 | 738 | 65 | |
GenBank mRNA | AB794093.1 | - | 8 - 9 | 649 | 65 | |
AB794094.1 | - | 7 - 8 | 598 | 65 | ||
AB794095.1 | - | 8 - 9 | 649 | 65 | ||
AB794096.1 | - | 8 - 9 | 649 | 65 | ||
AB794097.1 | - | 7 - 8 | 598 | 65 | ||
AB794098.1 | - | 7 - 8 | 598 | 65 | ||
AB794099.1 | - | 7 - 8 | 598 | 65 | ||
AB794100.1 | - | 7 - 8 | 598 | 65 | ||
AB794101.1 | - | 8 - 9 | 649 | 65 | ||
AB794102.1 | - | 7 - 8 | 585 | 65 | ||
AB794103.1 | - | 7 - 8 | 598 | 65 | ||
AF288137.1 | - | 7 - 8 | 700 | 65 | ||
AF463504.1 | - | 8 - 9 | 649 | 65 | ||
BC112910.1 | - | 8 - 9 | 789 | 6515946 | 65 | |
CF171555.1 | - | 5 - 6 | 511 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
