Assay Details
Gene Symbol: Gpr155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190297.2 | NP_001177226.1 | 5 - 6 | 1433 | 83 | |
NM_001276443.1 | NP_001263372.1 | 5 - 6 | 1433 | 83 | ||
NM_001276444.1 | NP_001263373.1 | 5 - 6 | 1433 | 83 | ||
GenBank mRNA | AK089436.1 | - | 5 - 6 | 1351 | 83 | |
BC115698.1 | - | 5 - 6 | 1329 | 40060105 | 83 | |
HM161763.2 | - | 5 - 6 | 1410 | 83 | ||
HM161764.2 | - | 5 - 6 | 1410 | 83 | ||
HM161765.2 | - | 5 - 6 | 1410 | 83 | ||
HM161766.2 | - | 5 - 6 | 1410 | 83 | ||
KF358283.1 | - | 5 - 6 | 1410 | 83 | ||
KF358284.1 | - | 5 - 6 | 1410 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
