Assay Details
Gene Symbol: Ptprj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135657.1 | NP_001129129.1 | 6 - 7 | 1179 | 63 | |
NM_008982.5 | NP_033008.3 | 6 - 7 | 1463 | 63 | ||
GenBank mRNA | AK147318.1 | - | 6 - 7 | 1384 | 63 | |
AK147556.1 | - | 6 - 7 | 1384 | 63 | ||
AY038891.1 | - | 6 - 7 | 1281 | 63 | ||
AY039232.1 | - | 6 - 7 | 1281 | 63 | ||
D45212.1 | - | 6 - 7 | 1419 | 63 | ||
D83203.1 | - | 6 - 7 | 1180 | 63 | ||
DQ133576.1 | - | 6 - 7 | 1194 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
