Assay Details
Gene Symbol: Begain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163175.1 | NP_001156647.1 | 6 - 7 | 538 | 57 | |
GenBank mRNA | AK158915.1 | - | 6 - 7 | 539 | 57 | |
BC080282.1 | - | 4 - 5 | 331 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062437 | BC059879.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062437 | BC080282.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062437 | NM_001163175.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062437 | AK158915.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224655 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819110 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336125 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115168 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875977 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204709 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696678 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469903 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35205 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575833 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 844381 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346121 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919372 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290732 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650267 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735998 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 792230 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622515 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947346 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410835 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147228 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129273 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12891 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062437 | BC059879.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062437 | BC080282.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062437 | NM_001163175.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062437 | AK158915.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07746243 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02603819 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02603787 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02603763 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02603797 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02603763 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731656 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09867574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10311793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09806678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10161055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09823317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731615 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10395168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10300696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731685 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731597 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09806671 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10324591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09867595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731798 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10324577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09789014 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731657 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731772 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09823328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10355965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731575 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09823323 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10306501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10324588 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731596 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10324583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731695 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731570 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731775 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10218727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731673 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731557 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731592 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10135708 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731777 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731523 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731667 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731623 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731630 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731629 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731806 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10218733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731654 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731574 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731588 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09849567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731784 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731521 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28731785 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10402603 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402601 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402600 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402602 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402599 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10402598 | - | - |
Affymetrix Mouse Clariom S Array | PSR1200016417.mm.1 | TC1200002364.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200016417.mm.1 | NM_001163175.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016417.mm.1 | NM_001163175.1 | Refseq |
Murine Genome U74v2 Set | 138102_at | - | - |
Murine Genome U74v2 Set | 138102_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 562314 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213442 | - | - |
