Assay Details
Gene Symbol: Ubr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC157930.1 | - | 3 - 4 | 723 | 65 | |
BC172022.1 | - | 3 - 4 | 724 | 65 | ||
BC172146.1 | - | 23 - 24 | 3289 | 65 | ||
BQ571759.1 | - | 1 - 2 | 199 | 65 | ||
DQ924536.1 | - | 23 - 24 | 3599 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
