Assay Details
Gene Symbol: Ubr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC157930.1 | - | 4 - 5 | 807 | 89 | |
BC172022.1 | - | 4 - 5 | 808 | 89 | ||
BC172146.1 | - | 24 - 25 | 3373 | 89 | ||
BQ571759.1 | - | 2 - 3 | 283 | 89 | ||
DQ924536.1 | - | 24 - 25 | 3683 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
