Assay Details
Gene Symbol: Ubr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004314.1 | - | 1 - 2 | 78 | ||
AK050359.1 | - | 5 - 6 | 544 | 78 | ||
AK173334.1 | - | 3 - 4 | 285 | 78 | ||
BC085286.1 | - | 6 - 7 | 670 | 78 | ||
BC157930.1 | - | 13 - 14 | 1975 | 78 | ||
BC172022.1 | - | 13 - 14 | 1976 | 78 | ||
BC172146.1 | - | 33 - 34 | 4542 | 78 | ||
DQ924536.1 | - | 33 - 34 | 4839 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
