Assay Details
Gene Symbol: Tubgcp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146190.2 | NP_666302.2 | 17 - 18 | 2466 | 115 | |
GenBank mRNA | AK032031.1 | - | 17 - 18 | 2426 | 115 | |
AK044887.1 | - | 17 - 18 | 2450 | 115 | ||
AK051326.1 | - | 17 - 18 | 2461 | 115 | ||
AK134963.1 | - | 9 - 10 | 1584 | 115 | ||
AK151454.1 | - | 16 - 17 | 2235 | 115 | ||
AK152033.1 | - | 16 - 17 | 2235 | 115 | ||
AK153331.1 | - | 16 - 17 | 2235 | 115 | ||
AK153357.1 | - | 16 - 17 | 2235 | 115 | ||
AK160939.1 | - | 17 - 18 | 2380 | 115 | ||
AK161681.1 | - | 18 - 19 | 2372 | 115 | ||
AK173291.1 | - | 14 - 15 | 1843 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
