Assay Details
Gene Symbol: Tubgcp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146190.2 | NP_666302.2 | 9 - 10 | 965 | 94 | |
GenBank mRNA | AK032031.1 | - | 9 - 10 | 925 | 94 | |
AK044444.1 | - | 9 - 10 | 851 | 94 | ||
AK044887.1 | - | 9 - 10 | 949 | 94 | ||
AK051326.1 | - | 9 - 10 | 960 | 94 | ||
AK086158.1 | - | 9 - 10 | 925 | 94 | ||
AK134963.1 | - | 1 - 2 | 83 | 94 | ||
AK151454.1 | - | 9 - 10 | 923 | 94 | ||
AK152033.1 | - | 9 - 10 | 923 | 94 | ||
AK153331.1 | - | 9 - 10 | 923 | 94 | ||
AK153357.1 | - | 9 - 10 | 923 | 94 | ||
AK160939.1 | - | 9 - 10 | 879 | 94 | ||
AK161681.1 | - | 9 - 10 | 923 | 94 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK086158.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
