Assay Details
Gene Symbol: Mrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008625.2 | NP_032651.2 | 3 - 4 | 742 | 89 | |
GenBank mRNA | BC141338.1 | - | 3 - 4 | 694 | 9055950 | 89 |
Z11974.1 | - | 3 - 4 | 742 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
