Assay Details
Gene Symbol: Ptpra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163688.1 | NP_001157160.1 | 11 - 12 | 1428 | 68 | |
NM_008980.2 | NP_033006.2 | 12 - 13 | 1536 | 68 | ||
GenBank mRNA | AF332081.1 | - | 10 - 11 | 1141 | 68 | |
AF332082.1 | - | 10 - 11 | 1141 | 68 | ||
AJ431367.1 | - | 10 - 11 | 1141 | 68 | ||
AK031690.1 | - | 11 - 12 | 1427 | 68 | ||
AK165673.1 | - | 11 - 12 | 1426 | 68 | ||
BC050930.1 | - | 11 - 12 | 1410 | 6811172 | 68 | |
BC110683.1 | - | 11 - 12 | 1394 | 3965311 | 68 | |
M36033.1 | - | 12 - 13 | 1274 | 68 | ||
Z23054.1 | - | 2 - 3 | 178 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
