Assay Details
Gene Symbol: Tubgcp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290824.1 | NP_001277753.1 | 10 - 11 | 1289 | 74 | |
NM_153387.3 | NP_700436.1 | 10 - 11 | 1289 | 74 | ||
NR_077245.2 | 10 - 11 | 1289 | 74 | |||
NR_110985.1 | 10 - 11 | 1289 | 74 | |||
GenBank mRNA | AK034694.1 | - | 1 - 2 | 819 | 74 | |
AK038921.1 | - | 10 - 11 | 1248 | 74 | ||
AK045305.1 | - | 10 - 11 | 1182 | 74 | ||
AK046132.1 | - | 10 - 11 | 1219 | 74 | ||
AK050250.1 | - | 10 - 11 | 1290 | 74 | ||
AK051517.1 | - | 10 - 11 | 1190 | 74 | ||
AK161774.1 | - | 1 - 2 | 85 | 74 | ||
AK181112.1 | - | 2 - 3 | 127 | 74 | ||
BC029106.1 | - | 10 - 11 | 1253 | 3711922 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
