Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290413.1 | NP_001277342.1 | 1 - 2 | 78 | 78 | |
NM_009422.3 | NP_033448.2 | 1 - 2 | 78 | 78 | ||
GenBank mRNA | AK152174.1 | - | 1 - 2 | 46 | 78 | |
BB841262.1 | - | 1 - 2 | 78 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK152174.1, NM_001290413.1, NM_009422.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
