Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009422.3 | NP_033448.2 | 2 - 3 | 303 | 55 | |
GenBank mRNA | AK038136.1 | - | 2 - 3 | 252 | 55 | |
AK052934.1 | - | 2 - 3 | 252 | 55 | ||
AK151653.1 | - | 2 - 3 | 241 | 55 | ||
AK152174.1 | - | 2 - 3 | 271 | 55 | ||
AK159581.1 | - | 2 - 3 | 273 | 55 | ||
AK159585.1 | - | 250 | 55 | |||
AK184085.1 | - | 1 - 2 | 56 | |||
AK184736.1 | - | 1 - 2 | 40 | 55 | ||
BB841262.1 | - | 2 - 3 | 303 | 55 | ||
BC003801.1 | - | 1 - 2 | 229 | 3492839 | 55 | |
BC060625.1 | - | 1 - 2 | 280 | 55 | ||
L35303.1 | - | 2 - 3 | 240 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
