Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290413.1 | NP_001277342.1 | 9 - 10 | 1269 | 83 | |
NM_009422.3 | NP_033448.2 | 9 - 10 | 1248 | 83 | ||
GenBank mRNA | AF027570.1 | - | 8 - 9 | 1160 | 83 | |
AK038136.1 | - | 9 - 10 | 1197 | 83 | ||
AK052934.1 | - | 9 - 10 | 1197 | 83 | ||
AK151653.1 | - | 9 - 10 | 1186 | 83 | ||
AK152174.1 | - | 9 - 10 | 1216 | 83 | ||
AK159581.1 | - | 9 - 10 | 1218 | 83 | ||
AK159585.1 | - | 1195 | 83 | |||
BC060625.1 | - | 7 - 8 | 3205 | 83 | ||
L35303.1 | - | 9 - 10 | 1185 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
